Orders LinkContactUsLinkComponent
Cryo gloves, waterproof
Cryo gloves, waterproof
Cryo gloves, waterproof
Certificates
Cryo gloves, waterproof
2 Options Available Below
Product Details & Documents
These waterproof cryo gloves are ideal for the safe handling of samples from ultra low and cryogenic storage.
  • Provides splash protection
  • Comfortable, lightweight, breathable
  • Side release buckles for quick on/off
Documents
Specifications for Product Family
Specifications
  • Catalog No:
  • THBR189441
  • THBR189442
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Cuff style:
  • Gauntlet
  • Gauntlet
Specifications
Product Family Options
Product Information
  • Model
    Glove size
    Colour
    Length
    Availability
    Price
  • Mid-arm length
    S
    Blue
    356 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 606,00 €
    606,00 €
    1 * 1 PAAR
       606,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    THBR189441
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Cuff style
    Gauntlet
  • Mid-arm length
    M
    Blue
    356 mm
    Log in for availability
    Log in for availability
    1 * 1 PAAR
    1 * 1 PAAR - 649,00 €
    649,00 €
    1 * 1 PAAR
       649,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    THBR189442
    Ambidextrous / hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Cuff style
    Gauntlet