Order Entry
Austria
Orders LinkContactUsLinkComponent
Deionizer, barnstead Bantam™

Deionizer, barnstead Bantam™

Bantam™ deioniser, 240 V
Catalog # BARND0805
Deionizer, barnstead Bantam™
Catalog # BARND0805
Deionizer, barnstead Bantam™
Deionizer, barnstead Bantam™

Bantam™ deioniser, 240 V

Price
1 600,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

It is a simple, cost effective method for deionisation or pretreatment of lab water. Typically these units are ideal where water usage is up to 38 litres per day.

Specifications
  • Description:
    Bantam™ deioniser, 240 V
  • Cat. No.:
    BARND0805
  • Capacity:
    38 L/day
  • Model:
    Bantam™
  • W×D×H:
    220×150×720 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.