Container, wide mouth, with hinged cap, Sterilin™
: Sterilin
Ideal for industrial sample collection, transport, storage and analysis.
- Aseptically manufactured
- Autoclavable at 121 °C with lid open
- Frosted writing area for sample identification
- Designed to stay vertical when opened
- Available in natural and the food industry standard of blue
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Frequently Bought Together