ContactUsLinkComponent
Tamper evident closures for PET/PETG square media bottles, HDPE
Tamper evident closures for PET/PETG square media bottles, HDPE
Catalog # NALG342141-0384
   Low Manufacturing Impact
undefined
Tamper evident closures for PET/PETG square media bottles, HDPE
Catalog # NALG342141-0384
   Low Manufacturing Impact
Specifications
  • For:
    Nalgene PET/PETG square media bottles (except 125 ml PETG media bottles)
  • Colour:
    Natural
  • Description:
    HDPE tamper evident closure
  • Material:
    HDPE
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Pk:
    240
  • Environmental certification:
    NAR Certificate of Retirement
Specifications
About this item
Closure, tamper evident, specially designed for use with PET/PETG square media bottles.
  • Natural HDPE closures are ADCF (Animal Derived Component-Free)
  • Fits Nalgene PET and PETG Media Bottles with 38-430 neck finish and leakproof guarantee
  • Easy application and breaking of the tamper evident seal ring
  • Gamma irradiated to provide 10⁻⁶ Sterility Assurance Level (SAL)
  • Flat top surface for possible printing or labelling
Packaging: Closures are sterile and bulk packed.