Order Entry
Austria
Orders LinkContactUsLinkComponent
Storage Boxes for Blocks

Storage Boxes for Blocks

Storage box, unfolded, White, 360×220×41 mm
Catalog # KLIN1606
Supplier:   KLINIPATH
Storage Boxes for Blocks
Catalog # KLIN1606
Supplier:  KLINIPATH
Storage Boxes for Blocks

Storage box, unfolded, White, 360×220×41 mm

Price
196,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

This storage box is complete with base and lid with writing area.

  • Made of white cardboard
  • Capacity approx. 250 blocks

The box is delivered without divider but with cap.

Specifications
  • Colour:
    White
  • Description:
    Storage box, unfolded
  • Seq:
    01
  • W×D×H:
    360×220×41 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.