Order Entry
Austria
ContactUsLinkComponent
Water purification systems, Barnstead™ LabTower™ TII
Water purification systems, Barnstead™ LabTower™ TII
Catalog # BARN50132141
undefined
Water purification systems, Barnstead™ LabTower™ TII
Catalog # BARN50132141

Specifications

  • Flow rate:
    1,5 l/min
  • Description:
    Barnstead™ LabTower™ TII 40 water purification systems, with UV lamp
  • Seq:
    04
  • Model:
    LabTower TII 40
  • Bacteria:
    <1 CFU/ml
  • Conductivity:
    0,1 - 1 µS/cm
  • Feed water:
    Tap water
  • Resistivity at +25 °C:
    10 - 1 MΏ.cm
  • W×D×H:
    450×580×1500 mm
  • Pk:
    1

Specifications

About this item

These water purification systems convert tap water into high purity water (Type II) and provide an integrated storage solution. Designed to continuously recirculate purified reservoir water to preserve water quality even during periods of inactivity.

  • Three system options are available with permeate flows of 20, 40 or 60 L/hr
  • Stand-alone design with integrated 100 L reservoir takes up zero bench space
  • 5 μm prefilter and activated carbon in the combi-filter cartridge protect the RO membrane from free chlorine and particles
  • Hardness stabilising cartridge protects the reverse osmosis stage from hard water
  • Microprocessor controlled for automatic operation with continuous monitoring of all critical parameters
  • Irradiation with UV light (TII units with UV option only) eliminates any bacteria or germs that are present
  • RS232 interface with adjustable send-interval for safe data transfer of all measured data to a PC computer or log printer

Typical applications: Rinsing lab glassware, supplying general lab equipment, preparing and diluting buffers, reagents and media.

Two ways to dispense water:
- Users can dispense directly from the system via the dispensing valve for calibration solutions, reagent preparation, filling of containers, general rinsing purposes, etc.
- Users can sispense from the reservoir to supply autoclaves, dishwashers, ultrapure water systems, etc.

Delivery: All systems include an RO membrane, a high purity water cartridge, 10" 5 μm prefilter with hardness stabiliser cartridge, pressure regulator, 0,2 µm filter, integrated 100 L reservoir and UV lamp where applicable.