Calcium perchlorate tetrahydrate, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15627-86-8
:
Formula:CaClO₄·4H₂O
MDL Number:MFCD00191895
Storage Temperature:Ambient
UN:1455
ADR:5.1,II
:GHS07
:GHS03
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Titration Complexometric:12.2 to 13.5 % (Calcium)
Specification Test Results
Frequently Bought Together