Order Entry
Austria
ContactUsLinkComponent
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Flammable
Irritant
Health
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Certificates
undefined
Flammable
Irritant
Health
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Certificates

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.05 % (Coulometric)
  • Density:
    (20°C) 1.032 to 1.036 g/ml
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    5 to 30 ppm BHT
  • Residue after evaporation:
    ≤0.005 %
  • Carbonyl (as HCHO):
    ≤0.01 %
  • Titrable acid:
    ≤0.0016 meq/g
  • Peroxides:
    ≤0.005 % (H2O2)
  • Freezing point:
    ≥11.0°C

Specification Test Results

Related Information

Frequently Bought Together