Sodium borohydride 12% in sodium hydroxide solution 40%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium tetrahydridoborate, Sodium tetrahydroborate
CAS Number:16940-66-2
:
Formula:NaBH₄
MDL Number:MFCD00003518
Molecular Weight:37.83 g/mol
Melting Pt:400
Flash Pt:69
ADR:8 II
Merck Index:13,08664
UN:3320
:GHS07
:GHS05
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless Liquid
- Titration Iodometric:11 to 13 %
Specification Test Results
Frequently Bought Together