



2,2,3-Trimethylbutane 99%
Catalog # ACRO398620050
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3880 to 1.3900 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
About this item
:
:100.2 g/mol
Boiling Pt:81 °C (1013 hPa)
Melting Pt:–25 °C
Density:0.69 g/cm³
Flash Pt:–7 °C
Storage Temperature:Ambient
UN:1206
ADR:3,II
:GHS07
:GHS08
:GHS09
:GHS02
CAS Number:464-06-2
Formula:C₇H₁₆
MDL Number:MFCD00039846