Order Entry
Austria
ContactUsLinkComponent
Sodium nitrite 99% contains an anticaking reagent, extra pure
Toxic
Environment
Oxidizing
Sodium nitrite 99% contains an anticaking reagent, extra pure
Certificates
undefined
Toxic
Environment
Oxidizing
Sodium nitrite 99% contains an anticaking reagent, extra pure

Specification Test Results

  • Appearance:
    Light yellow Crystalline powder
  • Water:
    ≤0.3 %
  • Titration:
    ≥98.5 %
  • Anticaking agent:
    ≤0.1 %

Specification Test Results

Related Information

Frequently Bought Together