
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate 99%
Catalog # ACRO363520010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Yellow to green Powder or crystals
- Infrared spectrum:Conforms
- Elemental Analysis:≥98.5 %
Specification Test Results
About this item
:
:444.239 g/mol
Melting Pt:230 °C
:GHS07
Storage Temperature:Ambient
CAS Number:21797-13-7
Formula:C₈H₁₂B₂F₈N₄Pd
MDL Number:MFCD00043297