Order Entry
Austria
ContactUsLinkComponent
 
Starch, soluble 1% (w/w) in aqueous solution stabilised, pure
 
undefined
Starch, soluble 1% (w/w) in aqueous solution stabilised, pure
 

 

  • Appearance:
    Clear to slightly hazy solution
  • Sensitivity:
    Passes test

 

 

Frequently Bought Together