
2-Methyl-3-phenylpropanoic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1009-67-2
:
Formula:C₁₀H₁₂O₂
MDL Number:MFCD00192301
:164.204 g/mol
Boiling Pt:167…168 °C (23 mmHg)
Melting Pt:37…41 °C
Flash Pt:113 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to light yellow Powder or crystals or crystalline mass and/or chunks
- Infrared spectrum:Conforms
- Melting point:37 to 41 °C
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together