Phosphorus pentoxide - methanesulphonic acid
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Eaton's Reagent
CAS Number:39394-84-8
:
MDL Number:MFCD00070545
:141.95 g/mol
Boiling Pt:122 °C (1 mmHg)
Density:1.5 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2922
ADR:8,II
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:to light brown or pale pink Solution (May contain precipitate, re-dissolves on warming)
- Refractive index:1.4325 to 1.4375 (20°C, 589 nm)
- Titration with NaOH:6.5 to 10.0 % (P2O5)
Specification Test Results
Frequently Bought Together