
Triphenylphosphine 1.0 - 1.5 mmol/g on resin, cross-linked with 1% DVB
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:39319-11-4
:
Formula:C₁₈H₁₅P
MDL Number:MFCD20489348
:262.29 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to yellow Beads
- Particle size:100 to 200 mesh
- Crosslinking:1 % DVB
- Loading:1 to 1.5 mmol/g
Specification Test Results
Frequently Bought Together