
Bis(acetonitrile)palladium(II) dichloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14592-56-4
:
Formula:C₄H₆Cl₂N₂Pd
MDL Number:MFCD00013122
:259.43 g/mol
Melting Pt:300 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:Orange to rust brown Crystalline powder
- Infrared spectrum:Conforms
- Elemental Analysis:≥98.5 %
Specification Test Results
Frequently Bought Together