Order Entry
Austria
ContactUsLinkComponent
(-)-Myrtenal 98%
(-)-Myrtenal 98%
Certificates
undefined
(-)-Myrtenal 98%

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5010 to 1.5050 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Specific optical rotation:
    -13° to -17° (20°C, 589 nm) (neat)

Specification Test Results

Related Information

Frequently Bought Together