
Levamisole hydrochloride 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16595-80-5
:
Formula:C₁₁H₁₂N₂S·HCl
MDL Number:MFCD00012675
:240.75 g/mol
Melting Pt:226…231 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤0.5 % (1 g, 105°C, 4 hrs)
- Specific optical rotation:-121° to -128° (20°C, 589 nm) (c=5, H2O) (on dried substance)
- Titration with NaOH:≥99.0 % (On dried substance)
Specification Test Results
Frequently Bought Together