Order Entry
Austria
ContactUsLinkComponent
Polyethyleneimine, M.W. ∼ 60,000 50% (w/w) in aqueous solution
Environment
Polyethyleneimine, M.W. ∼ 60,000 50% (w/w) in aqueous solution
Certificates
undefined
Environment
Polyethyleneimine, M.W. ∼ 60,000 50% (w/w) in aqueous solution

Specification Test Results

  • Appearance:
    Clear viscous liquid
  • Infrared spectrum:
    Conforms
  • Assay:
    48 to 52 % Solids
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together