Cellulose acetate ∼ M.W. 100,000
:9004-35-7
:
:C₂H₄O₂·(C₆H₁₀O₅)n
:MFCD00081496
:240 °C
:1.31 g/cm³
:Ambient
- Appearance:White Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5 %
- Acetyl content:39.8 % (typical)
- Hydroxyl content:3.6 % (typical)
Frequently Bought Together