Order Entry
Austria
ContactUsLinkComponent
 
pH Indicator solution universal in isopropanol (IPA), pure
  ACRO422435000
 :  
undefined
pH Indicator solution universal in isopropanol (IPA), pure
  ACRO422435000
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Appearance:
    Solution
  • Visual transition interval:
    pH 4.0 (red)

 

 

 :
 :19 °C
 :Ambient
 :1993
 :3,II
 :GHS02
 :GHS07
 :MFCD00148741