

Acrylamide : Bisacrylamide (19:1) 40% mix in water for biochemistry
:79-06-1
:MFCD00080848
:6.1,III
:GHS07
:GHS08
:Refrigerator
- Appearance:Clear colorless Solution
- UV:(1 % in water)
- Acrylic acid (HPLC):≤0.001 %
- Conductivity:≤3 µS/cm
Frequently Bought Together