Order Entry
Austria
ContactUsLinkComponent
 
Tris EDTA buffer, for molecular biology, DNAse, RNAse, protease free ready to use, pH 8,0
  ACRO327345000
 :  
undefined
Tris EDTA buffer, for molecular biology, DNAse, RNAse, protease free ready to use, pH 8,0
  ACRO327345000
 :  

 

  • Pk:
    500 mL
  • Description:
    Tris EDTA buffer, for molecular biology, DNAse, RNAse, Protease-free, ready to use, pH 8.0

 

 

CAS No.: 38641-82-6

MDL No.: MFCD00236359
Appearance: Clear colourless solution
Storage temperature: Ambient