Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solution pH 5, Acetate Buffer

Buffer solution pH 5, Acetate Buffer

Acetate buffer, Contains 0.7 ml glacial acetic acid and 0.6 g potassium hydroxide per litre., 4,98 - 5,02 (20 °C), Plastic bottle, 1 L
Catalog # ACRO258620010
Buffer solution pH 5, Acetate Buffer
Catalog # ACRO258620010
CAS Number:  
Buffer solution pH 5, Acetate Buffer

Acetate buffer, Contains 0.7 ml glacial acetic acid and 0.6 g potassium hydroxide per litre., 4,98 - 5,02 (20 °C), Plastic bottle, 1 L

CAS Number:  
Price
51,30 €
Log in for availability
1 * 1 L
Product Details & Documents

Documents

Specifications

Specifications

  • Concentration:
    Contains 0.7 ml glacial acetic acid and 0.6 g potassium hydroxide per litre.
  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Description:
    Acetate buffer
  • pH value:
    4,98 - 5,02 (20 °C)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.