Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer solution pH 10, carbonate buffer

Buffer solution pH 10, carbonate buffer

Buffer solution pH 10, carbonate buffer
Certificates
Buffer solution pH 10, carbonate buffer
Buffer solution pH 10, carbonate buffer

3 Options Available Below

Product Details & Documents

CAS No.: 1332-57-6

MDL No.: MFCD00036143
Appearance: Clear colourless solution
Storage Temperature: Ambient

Documents

Product Family Options

Product Information

  • pH value
    pH accuracy
    pH buffer type
    Colour
    Pack type
    Pk
    Availability
    Price
  • 10.00
    ±0,02
    Carbonate
    Colourless
    Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 59,80 €
    59,80 €
    1 * 1 L
       59,80 €

  • 10.00
    ±0,02
    Carbonate
    Colourless
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 37,90 €
    37,90 €
    1 * 500 mL
       37,90 €

  • 10.00
    ±0,02
    Carbonate
    Colourless
    Plastic bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L - 123,00 €
    123,00 €
    1 * 2,5 L
       123,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.