Order Entry
Austria
ContactUsLinkComponent
Prostaglandin E2 98%
Irritant
Prostaglandin E2 98%
Catalog # ACRO233560100
CAS Number:  
undefined
Irritant
Prostaglandin E2 98%
Catalog # ACRO233560100
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

Specification Test Results

  • Appearance:
    White to cream to light yellow Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥97.5 %

Specification Test Results

About this item

CAS Number:363-24-6
 :
Formula:C₂₀H₃₂O₅
MDL Number:MFCD00077861
Melting Pt:66…68 °C
 :352.47 g/mol
 :GHS07
Storage Temperature:Freezer