Order Entry
Austria
Orders LinkContactUsLinkComponent
Sodium fluoride 99+% for analysis

Sodium fluoride 99+% for analysis

Toxic
Sodium fluoride 99+% for analysis
Certificates
Sodium fluoride 99+% for analysis

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:7681-49-4
  • Formula:NaF
  • MDL Number:MFCD00003524
  • Molecular Weight:41.99 g/mol
  • Boiling Pt:1700 °C (1013 hPa)
  • Melting Pt:993 °C
  • Density:2,8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:1690
  • ADR:6.1,III

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Powder
  • Loss on drying:
    ≤0.2 % (150°C)
  • Titration:
    ≥99.0 %
  • Chloride (Cl):
    ≤50 ppm
  • Sulfate (SO4):
    ≤1000 ppm
  • Sodium fluosilicate:
    ≤0.1 %
  • Iron (Fe):
    ≤50 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Free acid (HF):
    ≤0.1 %
  • Free alkali (NaOH):
    ≤0.1 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Plastic bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,10 €
    21,10 €
    1 * 25 g
       21,10 €

  • Plastic bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 73,80 €
    73,80 €
    1 * 500 g
       73,80 €

  • Plastic bottle
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 228,00 €
    228,00 €
    1 * 2,5 kg
       228,00 €

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.