

2-Propanol 99.5+%, extra pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-63-0
:
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674
:60.1 g/mol
Boiling Pt:81…83 °C (760 mmHg)
Melting Pt:–89.5 °C
Density:0.785 g/cm³
Flash Pt:12 °C
Storage Temperature:Ambient
UN:1219
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.2 %
- GC:≥99.50 %
- Color scale:≤10 APHA
- Non-volatile matter:≤0.001 %
Specification Test Results
Frequently Bought Together