Order Entry
Austria
ContactUsLinkComponent
Photoreactive dyes and reagents
Photoreactive dyes and reagents
Catalog # 92016.
Supplier:  Biotium
CAS Number:  
undefined
Photoreactive dyes and reagents
Catalog # 92016.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Host:
  • Description:
    INA
  • Environmentally Preferable:

Specifications

About this item

INA (5-iodonaphthyl-1-azide) is a lipophilic photoreactive probe, which has been used to selectively label membrane-embedded cysteine residues of proteins.