Ice pans and buckets, multi-purpose, Corning®
: Corning
Ethylene vinyl acetate (EVA) foam.
- Solid, one piece moulded, unbreakable
- Resistant to highly acidic and basic solutions, salts, alcohol, acetone, chloroform, phenol, etc
- Insulative properties and resistant to temperatures from −196 to 93 °C
Ideal for use with ice, dry ice, liquid nitrogen, saline solution and acetone.
Corning ice pans and buckets are multi-purpose pans and buckets that are a great non-toxic alternative to conventional ice pans.
EVA has little or no odor, is inert and used in biomedical engineering applications such as drug delivery devices. The multi-purpose containers are highly insulative, stackable, unbreakable, lightweight and leak-proof. Containers will not 'sweat' or deform and have a textured finish on the bottom to prevent slipping on the benchtop. Ideal for use with ice, dry ice (−78 ℃), liquid nitrogen (−196 ℃), alcohol or saline solutions, or warm solutions (+93 °C). Available in five sizes and seven vibrant colours.
- Catalog No:
- 479-2014
- 216-1947
- 479-2013
- 216-1946
- 216-1945
- 479-1999
- 216-1944
- 216-1954
- 216-1943
- 216-1953
- 216-1942
- 216-1952
- 216-1951
- 216-1950
- 479-1977
- 479-1990
- 479-1989
- 479-1988
- 479-2016
- 479-1987
- 479-2015
- 479-2017
- 479-1997
- 479-1982
- 479-1996
- 479-1981
- 479-1995
- 479-1980
- 479-1979
- 479-1994
- 479-1978
- 479-1993
- 479-1992
- 479-1991
- 479-2018
- 216-1941
- 479-1986
- 216-1949
- 479-1985
- 479-1984
- 216-1948
- 479-1983
- 479-1998
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
Frequently Bought Together