Order Entry
Austria
ContactUsLinkComponent
Recombinant protein L, peroxidase conjugated, Pierce™
Recombinant protein L, peroxidase conjugated, Pierce™
Catalog # PIER32420
CAS Number:  
Recombinant protein L, peroxidase conjugated, Pierce™
Catalog # PIER32420
CAS Number:  

Specifications

  • Pk:
    0,5 mg
  • Description:
    Recombinant Protein L, peroxidase conjugated
  • Seq:
    04

Specifications

About this item

Pierce™ Recombinant Peroxidase Conjugated Protein L can be used to probe and detect rabbit and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

  • Contains four immunoglobulin-binding domains per protein
  • Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
  • Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins.