Order Entry
Austria
ContactUsLinkComponent
Anti-CD33 Antibody
Anti-CD33 Antibody
  BSBTPB9932
undefined
Anti-CD33 Antibody
  BSBTPB9932

 

  • Antibody type:
    Primary
  • Antigen name:
    CD33 molecule
  • Antigen symbol:
    CD33
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    945
  • Antigen synonyms:
    SIGLEC-3|SIGLEC3|p67
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Myeloid cell surface antigen CD33(CD33) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: CD33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human