ContactUsLinkComponent
Phosphoprotein phosphate estimation assay Kit, Pierce™
Phosphoprotein phosphate estimation assay Kit, Pierce™
Catalog # PIER23270
CAS Number:  
undefined
Phosphoprotein phosphate estimation assay Kit, Pierce™
Catalog # PIER23270
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Phosphoprotein Phosphate Estimation Assay Kit, sufficient for 500 tube assays or 1920 microplate assays
Specifications
About this item
Pierce™ Phosphoprotein Phosphate Estimation Assay Kit enables classification and identification of proteins as phosphorylated (serine and threonine), as well as semi-quantitative assessment of the phosphorylation level.
  • Specific – measures phosphoserine (p-Ser) and phosphothreonine (p-Thr) only
  • Test tube and 96-well microplate protocols included and require less than 90 minutes to perform
  • Semi-quantitative – calculate moles of phosphate per mole of protein using the included phosvitin standard
  • Format is adaptable to development of specific assays for pure phosphoproteins when previously characterised standards are available
Applications include identification of proteins as containing phosphoserine or phosphothreonine phosphorylations; estimating the amount of pS and pT phosphorylation; developing specific quantitative assays for well-characterised phosphoproteins of interest.
Delivery: Complete kit includes Ammonium molybdate Solution, 25 ml; Malachite green solution, 75 ml; Phosvitin positive control, 1 mg; and TBS pack (makes 500 ml).