VWR® pH Buffer, Acetic Acid-Ammonium Acetate, pH 6,8, REAGENT
Artikelnr. 85325.260
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- pH buffer type:Acetate
- Pk:500 mL
- Description:Acetic acid-ammonium acetate buffer TS
- Seq:02
- pH value:6,8
Specifications
About this item
Ready to use test solution (TS).
- Produced according to Pharmacopoeia requirements
- Reduces time and expenses
- Independent, traceable and certified
- Conforms to USP 34
- Certificates of analysis and safety data sheets available