Order Entry
Austria
ContactUsLinkComponent
Affinity purification resin, immobilised p-Aminophenyl phosphoryl choline gel, Pierce™
Affinity purification resin, immobilised p-Aminophenyl phosphoryl choline gel, Pierce™
Catalog # PIER20307
CAS Number:  
undefined
Affinity purification resin, immobilised p-Aminophenyl phosphoryl choline gel, Pierce™
Catalog # PIER20307
CAS Number:  

Specifications

  • Pk:
    5 ml
  • Description:
    Pierce™ Immobilised p-Aminophenyl phosphoryl choline gel
  • Matrix:
    Crosslinked 6% beaded agarose
  • Ligand:
    p-aminophenyl phosphoryl choline
  • Binding capacity:
    >3 mg of human C-reactive protein/ml
  • Type:
    Liquid

Specifications

About this item

Pierce™ Immobilised p-Aminophenyl Phosphoryl Choline Gel enables purification of C-reactive protein (CRP) from plasma, ascites and other biological fluids.

  • Format: 50% resin slurry in 0,02% sodium azide
  • Binding condition: Tris or borate-buffered saline (pH 8 to 8,5) with 1 to 2 mM calcium chloride
  • Elution condition: Tris or borate-buffered saline (pH 8 to 8,5) with 2 mM EDTA
  • Reusable: resin can be regenerated and reused several times