Order Entry
Austria
ContactUsLinkComponent
TAE buffer (20X), Pierce™
TAE buffer (20X), Pierce™
Catalog # PIER28354
CAS Number:  
undefined
TAE buffer (20X), Pierce™
Catalog # PIER28354
CAS Number:  

Specifications

  • Pk:
    0,5 L
  • Description:
    Pierce™ TAE buffer, 20X
  • Seq:
    02
  • pH value:
    8,3

Specifications

About this item

20X TAE buffer is a space-saving stock solution that is ideal for casting and preparing Tris-Acetate EDTA running buffer used for agarose gel electrophoresis of nucleic acids.

When diluted to 1X with water, yields 00,4 M Tris, 0,04 M acetate, 1 mM EDTA, pH 8,2 to 8,4.