ContactUsLinkComponent
Glutathione-S-Transferase, Pierce™
Glutathione-S-Transferase, Pierce™
Catalog # PIER20237
CAS Number:  
undefined
Glutathione-S-Transferase, Pierce™
Catalog # PIER20237
CAS Number:  
Specifications
  • Pk:
    1 mg
  • Description:
    Glutathione-S-transferase (GST)
  • Seq:
    01
Specifications
About this item
A solution of glutathione-S-transferase (≥100 units/mg).