



Mercury(II) chloride in aqueous solution USP Test Solution (TS)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7487-94-7
:
Formula:HgCl₂
MDL Number:MFCD00011041
:271.5 g/mol
Merck Index:13,05901
Melting Pt:273 °C
Storage Temperature:Ambient
EINECS:0
UN:2922
ADR:8,III
:GHS06
:GHS05
:GHS08
:GHS09
Related Information
Frequently Bought Together