Order Entry
Austria
ContactUsLinkComponent
Pierce 3UV UV Lamp
Pierce 3UV UV Lamp
Catalog # PIER95035
undefined
Pierce 3UV UV Lamp
Catalog # PIER95035

Specifications

  • Description:
    UV lamp, 3UV, 230 V, 50 Hz, 394×76×114 mm
  • Output:
    8 W
  • Pk:
    1
  • Wavelength:
    254 nm, 302 nm, 365 nm

Specifications

About this item

This compact ultraviolet lamp is intended for longwave, midrange and shortwave UV applications (252 nm, 302 nm and 365 nm). The lamp's innovative design allows users to dial-in the required wavelength with just the turn of a knob. The lamp is compatible with photo-reactive crosslinking and label-transfer reagents, including the Photo-reactive Amino Acids for discovering in-cell protein interactions.

  • Three wavelengths, 254/302/365 nm UV, in one lamp
  • Selection of each wavelength is by an easy turn of the dial located on one end of the housing
  • The uniquely designed reflector surfaces mounted behind each tube provides maximum UV for fluorescence applications
  • Lightweight and ergonomically designed for hand or stationary use
  • Design allows the stationary use by standing the lamp on its end or side
Caution: For research use only. Not for use in diagnostic procedures.