ContactUsLinkComponent
Modified lowry protein assay kit, Pierce™
Modified lowry protein assay kit, Pierce™
Catalog # PIER23240
CAS Number:  
undefined
Modified lowry protein assay kit, Pierce™
Catalog # PIER23240
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Modified Lowry Protein Assay Kit, sufficient reagents to perform 480 standard tube assays or 2,400 microplate assays
Specifications
About this item
The Modified Lowry Protein Assay Kit is the preferred colorimetric assay for quantifying total protein concentration.
  • Ready-to-use reagent for the loyal Lowry method user
  • Preformulated cupric sulfate-tartrate reagent stable for one year at room temperature
  • Linear results from 1 to 1500 µg/ml for BSA
  • Adaptable to microplates
  • Less protein: protein variation than dye-binding methods
The sensitivity of the Modified Lowry Protein Assay is greatly enhanced over that of the biuret reagent. The working range of the method extends from 5 to 2000 mg/ml. Using cold Modified Lowry Protein Assay Reagent will result in low absorbance values.
Delivery: Kit includes Modified Lowry Protein Assay Reagent (480 ml), 2 N Folin-Ciocalteu Phenol Reagent (50 ml), albumin standard ampoules, 2 mg/ml, (10×1 ml).