Order Entry
Austria
Orders LinkContactUsLinkComponent
Lead(II) acetate USP Test Solution (TS)

Lead(II) acetate USP Test Solution (TS)

Lead(II) acetate USP Test Solution (TS)
Supplier:  VWR Chemicals
Certificates
Lead(II) acetate USP Test Solution (TS)
Lead(II) acetate USP Test Solution (TS)

2 Options Available Below

Product Details & Documents

Ready to use test solution (TS).

  • Produced according to Pharmacopoeia requirements
  • Reduces time and expenses
  • Independent, traceable and certified
  • Conforms to USP 34
  • Certificates of analysis and safety data sheets available
Synonyms:  Acetic acid Lead(II) salt
  • CAS Number:301-04-2
  • Formula:(H₃CCOO)₂Pb
  • MDL Number:MFCD00012452
  • Molecular Weight:325.29 g/mol
  • Merck Index:12,05411
  • Melting Pt:280 °C
  • EINECS:0
  • UN:1616
  • ADR:6.1,III
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.