ContactUsLinkComponent
Pierce™ Peroxidase Suppressor
Pierce™ Peroxidase Suppressor
Catalog # PIER35000
CAS Number:  
undefined
Pierce™ Peroxidase Suppressor
Catalog # PIER35000
CAS Number:  
Specifications
  • Pk:
    100 ml
  • Description:
    Peroxidase suppressor
Specifications
About this item
This peroxidase suppressor is a stable and easy-to-use inhibitor of endogenous peroxidase activity from cells and tissues.
  • Stable and ready-to-use
  • Effectively inhibits peroxidase activity
  • No interference with antigen-antibody interactions
Designed to reduce background signal in HRP-based immunohistochemistry applications.
Caution: Store at 4 °C upon receipt.