
Hydrochloric acid 1 N USP Test Solution (TS)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
ADR:8,III
:36.46 g/mol
EINECS:0
UN:1789
:GHS05
Storage Temperature:Ambient
Frequently Bought Together