Order Entry
Austria
ContactUsLinkComponent
 
Iron(III) chloride in aqueous solution USP Test Solution (TS)
Catalog # 85347.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Iron(III) chloride in aqueous solution USP Test Solution (TS)
Catalog # 85347.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :162.21 g/mol
Merck Index:13,04048
Melting Pt:260 °C
Storage Temperature:Ambient
EINECS:0
UN:2582
ADR:8,III
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005