Order Entry
Austria
ContactUsLinkComponent
Pierce™ Conjugate Purification Kit
Pierce™ Conjugate Purification Kit
Catalog # PIER44920
CAS Number:  
undefined
Pierce™ Conjugate Purification Kit
Catalog # PIER44920
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Conjugate purification kit, sufficient for 10 purifications, each of 0,5 - 50 mg of enzyme-conjugated antibody

Specifications

About this item

Pierce™ conjugate purification kit is designed to eliminate excess, unconjugated HRP or alkaline phosphatase enzyme from labeled antibodies (IgG) after performing antibody-enzyme conjugation reactions. The single-use column-kit is intended for purification of 0,5 to 50 mg of antibody conjugate dissolved in tris-buffered saline.

  • Procedure is designed for removal of HRP or AP enzymes from IgG antibodies (not for use with IgM, IgY, Fab fragments or streptavidin conjugates)
  • Removing unconjugated enzyme from antibody conjugates often reduces background and improves overal performance of immunoassays
Delivery: Kit includes IDA agarose column, 2 ml (1 column), polyacrylamide desalting column, 10 ml (1 column), Column Activator Buffer (25 ml), Elution Buffer (35 ml), 2× TBS pack (each makes 500 ml), PBS pack (makes 500 ml), and Activity Preservation Solution (1 ml).