ContactUsLinkComponent
Substrate for western blotting, 1-Step™ NBT/BCIP
Substrate for western blotting, 1-Step™ NBT/BCIP
Catalog # PIER34042
CAS Number:  
undefined
Substrate for western blotting, 1-Step™ NBT/BCIP
Catalog # PIER34042
CAS Number:  
Specifications
  • Pk:
    250 ml
  • Description:
    1-Step™ NBT/BCIP, ready to use
Specifications
About this item
The 1-Step NBT/BCIP is a one-component sensitive precipitating alkaline phosphatase substrate for immunoblotting, immunohistochemistry, and in situ hybridisation applications.