ContactUsLinkComponent
Quantitative peroxide assay kit (lipid), Pierce™
Quantitative peroxide assay kit (lipid), Pierce™
  PIER23285
 :  
undefined
Quantitative peroxide assay kit (lipid), Pierce™
  PIER23285
 :  
  • Pk:
    1 KIT
  • Description:
    Quantitative peroxide assay kit (Lipid), sufficient for 100 tube assays or 500 microplate assays with lipid-containing samples
Pierce™ Quantitative Peroxide Assay Kit (Lipid) detects and measures hydrogen peroxide levels in biological samples using an iron and xylenol orange* (XO) reagent.
  • Ready to use – removes the variability, difficulty and expense of formulating in-house
  • Ideal for measurement of peroxide accumulation in lipid-containing samples
This lipid-compatible peroxide assay kit detects peroxides based on oxidation of ferrous to ferric ion in the presence of xylenol orange. It can be used with lipid-containing samples without extracting the lipids. The assay is less sensitive, since it does not contain sorbitol. Most proteins do not interfere with this assay, although some metal chelators may require use of a blank (ie. control) to mitigate these effects.
*Xylenol orange is (o-cresolsulfonephthalein-3´-3´-bis-[methyliminodiacetic acid sodium salt])
 : Complete kit includes Reagent A (1 ml) and Reagent C (4× 25 ml).
Reagents