VWR®, Melting point standards
Supplier: VWR Chemicals
Certificates
About this item
These standards can be used with any melting point apparatus and are prepared using pure raw materials. Melting points are determined using a high accuracy Differential Scanning Calorimeter (DSC) system that is calibrated to the ITS 90 International Temperature Scale.
- Ready to use
- Traceable
- Uncertainty of measurement up to ±0,3 °C
The melting points of these standards range from benzophenone (47 to 49 °C) to anthraquinone (283 to 286 °C).
Limited Time Sale
Select items below are currently on sale
Related Information
Certificates
Search by Lot NumberSDS_EN_CH_VWRC85110.pdf SDS_EN_DE_VWRC85101.pdf SDS_EN_DE_VWRC85110.pdf SDS_EN_NL_VWRC85110.pdf SDS_EN_UK_VWRC85101.pdf
SDS
More
spec_VWRC85322.001 spec_VWRC85322.001
Product specification
VWR® for Calibrations
Product Literature
Frequently Bought Together