Orders LinkContactUsLinkComponent
Trypsin protease, MS grade
Trypsin protease, MS grade
Trypsin protease, MS grade
Trypsin protease, MS grade
Pierce™ Trypsin Protease, MS Grade, is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.
  • Cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
  • No detectable chymotrypsin activity thus making it highly pure
  • Chemically modified for reduced autolytic activity
Applications include in-gel digestion of proteins from 1-D or 2-D gels and in-solution tryptic digestion of proteins. The selectivity of this enzyme is critical for reproducible protein digestion and mass spectrometry-based protein identification. It has been treated with TPCK to eliminate chymotryptic activity, improving digestion specificity. The enzyme is most active in pH ranges 7 to 9 and can be reversibly inactivated at pH <4.
Product Family Options
  • Description
    Pk
  • Trypsin Protease, 5×20 µg
    1 KIT
    1 * 1 KIT
    1 * 1 KIT - 111,00 €
    111,00 €
    1 * 1 KIT
       111,00 €
  • Trypsin Protease, 5×100 µg
    1 KIT
    1 * 1 KIT
    1 * 1 KIT - 479,00 €
    479,00 €
    1 * 1 KIT
       479,00 €
  • Trypsin Protease, 1 mg
    1 mg
    1 * 1 mg
    1 * 1 mg - 934,00 €
    934,00 €
    1 * 1 mg
       934,00 €
  • Trypsin Protease, frozen, 1×100 µg
    100 µG
    1 * 100 µG
    1 * 100 µG - 82,80 €
    82,80 €
    1 * 100 µG
       82,80 €